3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
CAS: 99-61-6 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA128411000
Price (MYR)
313.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
313.00
Add to cart
Specifications
Chemical Name or Material3-Nitrobenzaldehyde
Name Note99%
CAS99-61-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning