2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
CAS: 552-89-6 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA128400250
Price (MYR)
461.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
461.00
Add to cart
Chemical Identifiers
CAS3289-28-9
IUPAC Nameethyl cyclohexanecarboxylate
Molecular FormulaC9H16O2
InChI KeyJJOYCHKVKWDMEA-UHFFFAOYSA-N
SMILESCCOC(=O)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4395-1.4435 @ 20?C