4-Nitroaniline, 99%, Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
CAS: 100-01-6 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA128371000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7471-63-8
IUPAC Name7-methyl-8-nitroquinoline
Molecular FormulaC10H8N2O2
InChI KeyZZDTVYJYMRSNQL-UHFFFAOYSA-N
SMILESCC1=CC=C2C=CC=NC2=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Melting Point (clear melt)182-186?C
Appearance (Color)Cream to pale green-yellow or dark yellow to brown
Assay (HPLC)≥98.0%
Indicator
RUO – Research Use Only