Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 75-18-3 | C2H6S | 62.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
10 mL
1 L
Packaging:
Glass bottle
Catalog number FSA127760025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS598-94-7
IUPAC Name1,1-dimethylurea
Molecular FormulaC3H8N2O
InChI KeyYBBLOADPFWKNGS-UHFFFAOYSA-N
SMILESCN(C)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder