alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
alpha-Methylstyrene, 99%, stabilized, Thermo Scientific Chemicals
CAS: 98-83-9 | C9H10 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
Packaging:
Glass bottle
Catalog number FSA127710010
Price (MYR)
427.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
427.00
Add to cart
Chemical Identifiers
CAS7790-69-4
IUPAC Namelithium(1+) nitrate
Molecular FormulaLiNO3
InChI KeyIIPYXGDZVMZOAP-UHFFFAOYSA-N
SMILES[Li+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystalline powder
Titration after Ion exchange>=99.0 %
Appearance (Color)White
Loss on drying=<1 % (200°C)