p-Methylstyrene, 97%, stabilized, Thermo Scientific Chemicals
p-Methylstyrene, 97%, stabilized, Thermo Scientific Chemicals
p-Methylstyrene, 97%, stabilized, Thermo Scientific Chemicals
p-Methylstyrene, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Methylstyrene, 97%, stabilized, Thermo Scientific Chemicals
CAS: 622-97-9 | C9H10 | 118.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
Packaging:
Glass bottle
Catalog number FSA127700100
Price (MYR)
348.00
Add to cart
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
348.00
Add to cart
Chemical Identifiers
CAS21928-51-8
IUPAC Name5-bromo-1,2,3-trichlorobenzene
Molecular FormulaC6H2BrCl3
InChI KeyVZUMVBQMJFFYRM-UHFFFAOYSA-N
SMILESClC1=CC(Br)=CC(Cl)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%