1-Methyl-2-pyrrolidinone, 99%, extra pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99%, extra pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99%, extra pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99%, extra pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99%, extra pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99%, C5H9NO, CAS Number-872-50-4, 1-methyl-2-pyrrolidinone, 1-methyl-2-pyrrolidone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidinone, n-methylpyrrolidone, 2.5L
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
25 L
Packaging:
View all
Glass Bottle
Catalog number FSA127630025
Price (MYR)
738.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
738.00
Add to cart
Chemical Identifiers
CAS13817-37-3
IUPAC Nameλ²-cobalt(2+) tetrahydrate difluoride
Molecular FormulaCoF2H8O4
InChI KeyMNHUTFNEBYCZAF-UHFFFAOYSA-L
SMILESO.O.O.O.[F-].[F-].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Suitable for battery materials development.
RUO – Research Use Only