1-Methyl-2-pyrrolidinone, 99%, extra pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99%, extra pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99%, extra pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99%, extra pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99%, extra pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99%, C5H9NO, CAS Number-872-50-4, 1-methyl-2-pyrrolidinone, 1-methyl-2-pyrrolidone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidinone, n-methylpyrrolidone, 2.5L
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number FSA127630010
Price (MYR)
499.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
499.00
Add to cart
Chemical Identifiers
CAS25322-68-3
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.5 % (K.F.)
Infrared spectrumConforms
Hydroxyl value17 to 20 mg KOH/g
Appearance (Color)White
Appearance (Form)Powder or flakes
Suitable for battery materials development.
RUO – Research Use Only