Methyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl methacrylate, 99%, C5H8O2, CAS Number-80-62-6, 150-76-5, acryester m, diakon, methacrylic acid methyl ester, methyl 2-methyl-2-propenoate, methyl 2-methylpropenoate, methyl-methacrylat, methylmethacrylate, methyl methacrylate, methyl methylacrylate, pegalan, 1L
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA127140250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS350-46-9
IUPAC Name1-fluoro-4-nitrobenzene
Molecular FormulaC6H4FNO2
InChI KeyWFQDTOYDVUWQMS-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)> 98.5%
Formliquid as melt
Clarity25% in ethanol, clear solution
Refractive Index1.5300 - 1.5330 @20?C
Appearance (Color)Clear, yellow