2-Methylhexane, 99%, Thermo Scientific Chemicals
2-Methylhexane, 99%, Thermo Scientific Chemicals
2-Methylhexane, 99%, Thermo Scientific Chemicals
2-Methylhexane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methylhexane, 99%, Thermo Scientific Chemicals
CAS: 591-76-4 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA126910250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7553-56-2
IUPAC Namediiodine
Molecular FormulaI2
InChI KeyPNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILESII
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown
Titration with Na2S2O30.0998 to 0.1002 N (20°C)
Appearance (Form)Solution