Methyl formate, 97%, pure, Thermo Scientific Chemicals
Methyl formate, 97%, pure, Thermo Scientific Chemicals
Methyl formate, 97%, pure, Thermo Scientific Chemicals
Methyl formate, 97%, pure, Thermo Scientific Chemicals
Methyl formate, 97%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl formate, 97%, pure, Thermo Scientific Chemicals
CAS: 107-31-3 | C2H4O2 | 60.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA126820010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl formate
Name Note97%
CAS107-31-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Extremely flammable liquid and vapor.
View more
Suitable for battery materials development.
RUO – Research Use Only