2-Methylbutane, 99+%, extra pure, Thermo Scientific Chemicals
2-Methylbutane, 99+%, extra pure, Thermo Scientific Chemicals
2-Methylbutane, 99+%, extra pure, Thermo Scientific Chemicals
2-Methylbutane, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methylbutane, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 78-78-4 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number FSA126470010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS7440-65-5
IUPAC Nameyttrium
Molecular FormulaY
InChI KeyVWQVUPCCIRVNHF-UHFFFAOYSA-N
SMILES[Y]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.