Methyl 4-bromocrotonate, 85%, tech., Thermo Scientific Chemicals
Methyl 4-bromocrotonate, 85%, tech., Thermo Scientific Chemicals
Methyl 4-bromocrotonate, 85%, tech., Thermo Scientific Chemicals
Methyl 4-bromocrotonate, 85%, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl 4-bromocrotonate, 85%, tech., Thermo Scientific Chemicals
CAS: 1117-71-1 | C5H7BrO2 | 179.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Catalog number FSA126455000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight179.01
GradeTechnical