Methyl benzoate, 99%, Thermo Scientific Chemicals
Methyl benzoate, 99%, Thermo Scientific Chemicals
Methyl benzoate, 99%, Thermo Scientific Chemicals
Methyl benzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl benzoate, 99%, Thermo Scientific Chemicals
CAS: 93-58-3 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA126345000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS90-12-0
IUPAC Name1-methylnaphthalene
Molecular FormulaC11H10
InChI KeyQPUYECUOLPXSFR-UHFFFAOYSA-N
SMILESCC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=95 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow to light brown
Refractive index1.6100 to 1.6180 (20°C, 589 nm)