N-Methylaniline, 99%, Thermo Scientific Chemicals
N-Methylaniline, 99%, Thermo Scientific Chemicals
N-Methylaniline, 99%, Thermo Scientific Chemicals
N-Methylaniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Methylaniline, 99%, Thermo Scientific Chemicals
CAS: 100-61-8 | C7H9N | 107.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass bottle
Catalog number FSA126270010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112529-15-4
IUPAC Namehydrogen 5-({4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl}methyl)-1,3-thiazolidine-2,4-dione chloride
Molecular FormulaC19H21ClN2O3S
InChI KeyGHUUBYQTCDQWRA-UHFFFAOYNA-N
SMILES[H+].[Cl-].CCC1=CC=C(CCOC2=CC=C(CC3SC(=O)NC3=O)C=C2)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%