Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
CAS: 96-33-3 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
500 mL
1 L
2.5 L
Packaging:
Catalog number FSA126190100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS6131-90-4
IUPAC Namesodium acetate trihydrate
Molecular FormulaC2H9NaO5
InChI KeyAYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILESO.O.O.[Na+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO499.0 to 101.0 %
Heavy metals (as Pb)=<5 ppm
Potassium (K)=<0.005 %
Sulfate (SO4)=<0.002 %