Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
CAS: 96-33-3 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
10 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA126190010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS68855-54-9
SpecificationsSpecification SheetSpecification Sheet
CommentPermeability 0.889-1.488 Darcy
Particle Size+150 mesh ≤12.0%
Appearance (Color)White to pale cream or pale grey
FormPowder
CommentWet density <421.41 g/L