4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
CAS: 150-76-5 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
500 g
1 kg
Catalog number FSA126000051
Price (MYR)
3,064.00
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (MYR)
3,064.00
Add to cart
Specifications
Chemical Name or Material4-Methoxyphenol
Name NoteStabilized
CAS150-76-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning