4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
CAS: 150-76-5 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
5 kg
Catalog number FSA126000010
Price (MYR)
823.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
823.00
Add to cart
Chemical Identifiers
CAS1454-53-1
IUPAC Nameethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
Molecular FormulaC15H20ClNO3
InChI KeyYPFMNHZRNXPYBG-UHFFFAOYNA-N
SMILESCl.CCOC(=O)C1CN(CC2=CC=CC=C2)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pink
Infrared spectrumConforms
Titration Argentometric>=96.0 % (on dry substance)
Appearance (Form)Powder
Titration with HClO4>=96.0 % (On dry substance)