p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Methoxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
CAS: 830-09-1 | C10H10O3 | 178.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA125840250
Price (MYR)
469.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
469.00
Add to cart
Chemical Identifiers
CAS5949-29-1
IUPAC Name2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
Molecular FormulaC6H10O8
InChI KeyYASYEJJMZJALEJ-UHFFFAOYSA-N
SMILESO.OC(=O)CC(O)(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH99.4 to 101.0 %