Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
CAS: 141-79-7 | C6H10O | 98.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA125600025
Price (MYR)
924.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
924.00
Add to cart
Chemical Identifiers
CAS77-95-2
IUPAC Name1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid
Molecular FormulaC7H12O6
InChI KeyAAWZDTNXLSGCEK-UHFFFAOYNA-N
SMILESOC1CC(O)(CC(O)C1O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point162°C to 168°C
Titration with NaOH>=98.0 %
Specific optical rotation-42° to -45° (20°C, 589 nm) (c=10, water)
Infrared spectrumConforms
Appearance (Color)White to light yellow