Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mesityl oxide, 99%, mixture of alpha- and beta-isomers, Thermo Scientific Chemicals
CAS: 141-79-7 | C6H10O | 98.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA125600010
Price (MYR)
496.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
496.00
Add to cart
Chemical Identifiers
CAS29124-57-0
IUPAC Name2-amino-5-bromobenzaldehyde
Molecular FormulaC7H6BrNO
InChI KeyVBYZWJMZASVGNB-UHFFFAOYSA-N
SMILESNC1=C(C=O)C=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Free acid (titration)≤1.5%
Melting Point (clear melt)74.0-80.0?C
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%