2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Mercapto-1-methylimidazole, 98%, Thermo Scientific Chemicals
2-Mercapto-1-methylimidazole, CAS # 60-56-0, also known as methimazole, is a thyroid hormone antagonist used to study the treatment of thyroid disorders.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA125500250
Price (MYR)
612.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
612.00
Add to cart
Specifications
Chemical Name or Material2-Mercapto-1-methylimidazole
CAS60-56-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Health Hazard 3GHS P Statement
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Contaminated work clothing should not be allowed out of the workplace.
View more
General Description
  • 2-Mercapto-1-methylimidazole, also known as methimazole, is an antithyroid medication
  • Methimazole works by inhibiting the enzyme thyroid peroxidase (TPO), which is responsible for synthesizing the thyroid hormones triiodothyronine (T3) and thyroxine (T4) from the precursor thyroglobulin
Applications
  • Methimazole has been used in laboratory settings to model olfactory epithelial degeneration
  • May be used to study disruptions in iodide metabolism
  • Methimazole has been used in Pharmacokinetics studies in the laboratory
RUO – Research Use Only
General References:
  1. Stanley, M.M.; Astwood, E.B. 1-Methyl-2-mercaptoimidazole; an antithyroid compound highly active in man. Endocrinology. 1949, 44(6), 588.
  2. Ogawa, T.; Takezawa, K.; Shimizu, S.; Shimizu, T. Valproic acid promotes neural regeneration of olfactory epithelium in adult mice after methimazole-induced damage. Am J Rhinol Allergy. 2014, 28(2), e95-9.
  3. Furman, B.L. in xPharm: The Comprehensive Pharmacology Reference. Methimazole. 2007 pp 1-4.