Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
CAS: 68-11-1 | C2H4O2S | 92.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
10 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA125430025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS59-30-3
IUPAC Name(2S)-2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
Molecular FormulaC19H19N7O6
InChI KeyOVBPIULPVIDEAO-LBPRGKRZSA-N
SMILESNC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)=CN=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=96.0 %
Sulfated ash=<0.2 %