DL-Menthol, 99%, Thermo Scientific Chemicals
DL-Menthol, 99%, Thermo Scientific Chemicals
DL-Menthol, 99%, Thermo Scientific Chemicals
DL-Menthol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Menthol, 99%, Thermo Scientific Chemicals
CAS: 89-78-1 | C10H20O | 156.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
Packaging:
Glass bottle
Catalog number FSA125395000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDL-Menthol
Name Note99%
CAS89-78-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement