DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Mandelic acid, 99+%, Thermo Scientific Chemicals
CAS: 90-64-2 | C8H8O3 | 152.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
Packaging:
Plastic bottle
Catalog number FSA125311000
Price (MYR)
238.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
238.00
Add to cart
Chemical Identifiers
CAS107-10-8
IUPAC Namepropan-1-amine
Molecular FormulaC3H9N
InChI KeyWGYKZJWCGVVSQN-UHFFFAOYSA-N
SMILESCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 % (K.F.)
Refractive index1.3870 to 1.3890 (20°C, 589 nm)
Color scale=<20 APHA
Infrared spectrumConforms
GC>=99.0 %