Malonic acid, 99%, Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Malonic acid, 99%, Thermo Scientific Chemicals
CAS: 141-82-2 | C3H4O4 | 104.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA125262500
Price (MYR)
550.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
550.00
Add to cart
Chemical Identifiers
CAS143-33-9
IUPAC Namesodium iminomethanide
Molecular FormulaCNNa
InChI KeyMNWBNISUBARLIT-UHFFFAOYSA-N
SMILES[Na+].[C-]#N
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=97 %
Sulfate (SO4)=<0.025 %
Sulfide (S)=<0.003 %
Copper (Cu)=<0.002 %
Appearance (Color)White