DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
CAS: 6915-15-7 | C4H6O5 | 134.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA125252500
Price (MYR)
259.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
259.00
Add to cart
Specifications
Chemical Name or MaterialDL-Malic acid
CAS6915-15-7
Melting Point128.0°C to 132.0°C
Health Hazard 1Danger
Health Hazard 2GHS P Statement Causes serious eye irritation.