DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
CAS: 6915-15-7 | C4H6O5 | 134.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
500 g
1 kg
Packaging:
View all
Plastic Drum
Catalog number FSA125250050
Price (MYR)
1,123.00
EA
Add to cart
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
1,123.00
EA
Add to cart
Chemical Identifiers
CAS16940-66-2
IUPAC Namesodium boranuide
Molecular FormulaBH4Na
InChI KeyYOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES[BH4-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Form-4+40 Mesh Granules
Assay (unspecified)>97.0%