Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
25 g
1 kg
5 kg
Packaging:
View all
Fibre drum
Catalog number FSA125240251
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
Chemical Identifiers
CAS64-20-0
IUPAC Nametetramethylazanium bromide
Molecular FormulaC4H12BrN
InChI KeyDDFYFBUWEBINLX-UHFFFAOYSA-M
SMILES[Br-].C[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤1%