Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
25 g
1 kg
25 kg
Packaging:
View all
Plastic drum
Catalog number FSA125240050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS18282-10-5
IUPAC Namestannanedione
Molecular FormulaO2Sn
InChI KeyXOLBLPGZBRYERU-UHFFFAOYSA-N
SMILESO=[Sn]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.004% max.