Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Maleic anhydride, 99%, pastilles, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number FSA125240010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMaleic anhydride
CAS108-31-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes damage to organs through
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre