Maleic Acid, 99%, Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Maleic Acid, 99%, Thermo Scientific Chemicals
CAS: 110-16-7 | C4H4O4 | 116.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA125230010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS100-76-5
IUPAC Name1-azabicyclo[2.2.2]octane
Molecular FormulaC7H13N
InChI KeySBYHFKPVCBCYGV-UHFFFAOYSA-N
SMILESC1CN2CCC1CC2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
FormCrystals or crystalline powder or powder
Assay (Non-aqueous acid-base Titration)≥97.0 to ≤103.0%
CommentProduct may be sourced from UK or US
Loss on Drying≤1.0% (US sourced material)