L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
CAS: 79-33-4 | C3H6O3 | 90.078 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA125070010
Price (MYR)
389.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
389.00
Add to cart
Chemical Identifiers
CAS13735-13-2
IUPAC Name6-bromo-3,4-dihydro-2H-1-benzothiopyran-4-one
Molecular FormulaC9H7BrOS
InChI KeySBNPYDJTVGRDAA-UHFFFAOYSA-N
SMILESBrC1=CC2=C(SCCC2=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream
Melting Point (clear melt)66.5-75.5?C
FormPowder
Assay (GC)≥94.0%