DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
CAS: 50-21-5 | C3H6O3 | 90.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number FSA125065000
Price (MYR)
430.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
430.00
Add to cart
Chemical Identifiers
CAS4891-38-7
IUPAC Namemethyl 3-phenylprop-2-ynoate
Molecular FormulaC10H8O2
InChI KeyJFGWPXKGINUNDH-UHFFFAOYSA-N
SMILESCOC(=O)C#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.5595-1.5645 @ 20?C
FormLiquid
Assay (GC)≥96.0%