DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Lactic acid, 90%, Thermo Scientific Chemicals
CAS: 50-21-5 | C3H6O3 | 90.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number FSA125060010
Price (MYR)
608.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
608.00
Add to cart
Specifications
Chemical Name or MaterialDL-Lactic acid
CAS50-21-5
Melting Point18.0°C
Boiling Point122.0°C (15.0 mmHg)
Health Hazard 1Danger