Octadecylbenzene, 98%, Thermo Scientific Chemicals
Octadecylbenzene, 98%, Thermo Scientific Chemicals
Octadecylbenzene, 98%, Thermo Scientific Chemicals
Octadecylbenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Octadecylbenzene, 98%, Thermo Scientific Chemicals
CAS: 4445-07-2 | C24H42 | 330.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
50 g
Packaging:
Glass bottle
Catalog number FSA124700100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13324-11-3
IUPAC Namemethyl 2-chloro-4-nitrobenzoate
Molecular FormulaC8H6ClNO4
InChI KeyPICNSXCJRMYANX-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C=C1Cl)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)73.0-79.0?C
Appearance (Color)White to pale cream
Assay (GC)≥97.5%