Hydrochloric acid, for analysis, fuming, 37% solution in water
Hydrochloric acid, for analysis, fuming, 37% solution in water
Hydrochloric acid, for analysis, fuming, 37% solution in water
Hydrochloric acid, for analysis, fuming, 37% solution in water
Thermo Scientific Chemicals
Hydrochloric acid, for analysis, fuming, 37% solution in water
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA1246300111 LPlastic bottle
Catalog number FSA124630011
Price (MYR)
329.00
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
329.00
Add to cart
Specifications
Chemical Name or MaterialHydrochloric acid
CAS7647-01-0
Health Hazard 1Danger
Density1.1800g/mL
Health Hazard 2GHS P Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May be corrosive to metals.
View more
For analysis
RUO – Research Use Only