β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
β-Nicotinamide adenine dinucleotide hydrate, 98+%, Thermo Scientific Chemicals
CAS: 53-84-9 | C21H27N7O14P2 | 663.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Packaging:
Glass bottle
Catalog number FSA124530050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS86-55-5
IUPAC Namenaphthalene-1-carboxylic acid
Molecular FormulaC11H8O2
InChI KeyLNETULKMXZVUST-UHFFFAOYSA-N
SMILESOC(=O)C1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder
Appearance (Color)Light yellow to brown
Melting point160°C to 164°C