Sulfuric acid, 1N (0.5M) standard solution, Thermo Scientific Chemicals
Sulfuric acid, 1N (0.5M) standard solution, Thermo Scientific Chemicals
Sulfuric acid, 1N (0.5M) standard solution, Thermo Scientific Chemicals
Sulfuric acid, 1N (0.5M) standard solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfuric acid, 1N (0.5M) standard solution, Thermo Scientific Chemicals
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic bottle
Catalog number FSA124240025
Price (MYR)
629.00
Add to cart
Quantity:
2.5 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
629.00
Add to cart
Chemical Identifiers
CAS31252-42-3
IUPAC Name4-benzylpiperidine
Molecular FormulaC12H17N
InChI KeyABGXADJDTPFFSZ-UHFFFAOYSA-N
SMILESC(C1CCNCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=97.5 %
Appearance (Color)Clear colorless to yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=97.5 %
View more
Volumetric standard solution
RUO – Research Use Only