Sulfuric acid, 1N (0.5M) standard solution, Thermo Scientific Chemicals
Sulfuric acid, 1N (0.5M) standard solution, Thermo Scientific Chemicals
Sulfuric acid, 1N (0.5M) standard solution, Thermo Scientific Chemicals
Sulfuric acid, 1N (0.5M) standard solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfuric acid, 1N (0.5M) standard solution, Thermo Scientific Chemicals
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic bottle
Catalog number FSA124240010
Price (MYR)
386.00
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
386.00
Add to cart
Specifications
Chemical Name or MaterialSulfuric acid
ColorColorless
Name Note1N (0.5M) Standard Solution
CAS7664-93-9
Concentration0.998 to 1.002N (20°C)
View more
Volumetric standard solution
RUO – Research Use Only