Sulfuric acid, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals
Sulfuric acid, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals
Sulfuric acid, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals
Sulfuric acid, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfuric acid, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic Bottle
Catalog number FSA124230025
Price (MYR)
456.00
Add to cart
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
456.00
Add to cart
Chemical Identifiers
CAS20859-02-3
IUPAC Name(2S)-2-amino-3,3-dimethylbutanoic acid
Molecular FormulaC6H13NO2
InChI KeyNPDBDJFLKKQMCM-SGAVLPGINA-N
SMILESCC(C)(C)[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Optical Rotation+7.0? ± 1.0? (c=5 in 5N HCl)
Volumetric standard solution
RUO – Research Use Only