Sulfuric acid, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals
Sulfuric acid, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals
Sulfuric acid, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals
Sulfuric acid, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfuric acid, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
Catalog number FSA124230010
Price (MYR)
357.00
Add to cart
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
357.00
Add to cart
Chemical Identifiers
CAS1733-12-6
IUPAC Name3,3-bis(4-hydroxy-3-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC21H18O5S
InChI KeyOBRMNDMBJQTZHV-UHFFFAOYSA-N
SMILESCC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific extinction E (1%/1cm)>=1500 (572 nm, in 0.01 N NaOH)
Appearance (Color)Dark green to red-brown
Infrared spectrumConforms
Solubility(0.1 % in 50 % ethanol) Clear
Visual transition intervalfrom pH 7.0 (orange) to pH 8.8 (purple)
View more
Volumetric standard solution
RUO – Research Use Only