Dichloromethane, 99+%, extra pure, stabilized with ethanol, Thermo Scientific Chemicals
Dichloromethane, 99+%, extra pure, stabilized with ethanol, Thermo Scientific Chemicals
Dichloromethane, 99+%, extra pure, stabilized with ethanol, Thermo Scientific Chemicals
Dichloromethane, 99+%, extra pure, stabilized with ethanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloromethane, 99+%, extra pure, stabilized with ethanol, Thermo Scientific Chemicals
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA124050025
Price (MYR)
402.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
402.00
Add to cart
Chemical Identifiers
CAS108-32-7
IUPAC Name4-methyl-1,3-dioxolan-2-one
Molecular FormulaC4H6O3
InChI KeyRUOJZAUFBMNUDX-UHFFFAOYNA-N
SMILESCC1COC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<40 APHA
Infrared spectrumConforms
GC>=99.4 %
Water=<0.2 %