4-Chlorophenylacetyl chloride, 96%, Thermo Scientific Chemicals
4-Chlorophenylacetyl chloride, 96%, Thermo Scientific Chemicals
4-Chlorophenylacetyl chloride, 96%, Thermo Scientific Chemicals
4-Chlorophenylacetyl chloride, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorophenylacetyl chloride, 96%, Thermo Scientific Chemicals
CAS: 25026-34-0 | C8H6Cl2O | 189.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA123450050
Price (MYR)
236.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
236.00
Add to cart
Chemical Identifiers
CAS92-51-3
IUPAC Name1,1'-bi(cyclohexane)
Molecular FormulaC12H22
InChI KeyWVIIMZNLDWSIRH-UHFFFAOYSA-N
SMILESC1CCC(CC1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4780 to 1.4800 (20°C, 589 nm)