Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2 kg
Packaging:
View all
Glass bottle
Catalog number FSA122650050
Price (MYR)
218.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
218.00
Add to cart
Specifications
Chemical Name or MaterialIsophthalic acid
CAS121-91-5
Melting Point341.0°C to 343.0°C
ColorWhite
EINECS Number204-506-4