Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isophthalic acid, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number FSA122650020
Price (MYR)
777.00
Add to cart
Quantity:
2 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
777.00
Add to cart
Chemical Identifiers
CAS6797-79-1
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6045-1.6085 @ 20?C
Appearance (Color)Clear colorless to pale red to red to brown
FormLiquid
Assay (GC)≥97.5%