Isonipecotic Acid, 99%
Isonipecotic Acid, 99%
Isonipecotic Acid, 99%
Isonipecotic Acid, 99%
Thermo Scientific Chemicals
Isonipecotic Acid, 99%
CAS: 498-94-2 | C6H11NO2 | 129.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA122620250
Price (MYR)
369.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
369.00
Add to cart
Chemical Identifiers
CAS144-48-9
IUPAC Name2-iodoacetamide
Molecular FormulaC2H4INO
InChI KeyPGLTVOMIXTUURA-UHFFFAOYSA-N
SMILESNC(=O)CI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point>=90°C
Titration Argentometric>=97.5 % (ex I)
Appearance (Color)White to yellow to beige