Isonicotinic acid hydrazide, 99%, Thermo Scientific Chemicals
Isonicotinic acid hydrazide, 99%, Thermo Scientific Chemicals
Isonicotinic acid hydrazide, 99%, Thermo Scientific Chemicals
Isonicotinic acid hydrazide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isonicotinic acid hydrazide, 99%, Thermo Scientific Chemicals
CAS: 54-85-3 | C6H7N3O | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number FSA122600050
Price (MYR)
254.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
254.00
Add to cart
Chemical Identifiers
CAS12055-23-1
IUPAC Namehafnium(4+) dioxidandiide
Molecular FormulaHfO2
InChI KeyWIHZLLGSGQNAGK-UHFFFAOYSA-N
SMILES[O--].[O--].[Hf+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity content0.2% Zr max.
Total Metal Impurities excluding Zr0.005% max.