2-Iodotoluene, 98%
2-Iodotoluene, 98%
2-Iodotoluene, 98%
Thermo Scientific Chemicals
2-Iodotoluene, 98%
CAS: 615-37-2 | C7H7I | 218.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA12243025025 mLGlass bottle
Catalog number FSA122430250
Price (MYR)
624.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
624.00
Add to cart
Chemical Identifiers
CAS100-76-5
IUPAC Name1-azabicyclo[2.2.2]octane
Molecular FormulaC7H13N
InChI KeySBYHFKPVCBCYGV-UHFFFAOYSA-N
SMILESC1CN2CCC1CC2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
FormCrystals or crystalline powder or powder
Assay (Non-aqueous acid-base Titration)≥97.0 to ≤103.0%
CommentProduct may be sourced from UK or US
Loss on Drying≤1.0% (US sourced material)